Assay Details
| Gene Symbol: GPX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046088.2 | NP_001039553.1 | 2 - 3 | 485 | 128 | |
| GenBank mRNA | BC111287.1 | - | 2 - 3 | 485 | 128 | |
More Information
Set Membership: |
|

GPX8
Genomic Map