Assay Details
| Gene Symbol: ANKRD61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077072.1 | NP_001070540.1 | 2 - 3 | 426 | 86 | |
| GenBank mRNA | BC111230.1 | - | 2 - 3 | 426 | 86 | |
More Information
Set Membership: |
|

ANKRD61
Genomic Map