Assay Details
| Gene Symbol: FAM136A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046588.2 | NP_001040053.1 | 1 - 2 | 126 | 83 | |
| GenBank mRNA | BC105349.1 | - | 1 - 2 | 128 | 83 | |
More Information
Set Membership: |
|

FAM136A
Genomic Map