Assay Details
| Gene Symbol: BRMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076404.1 | NP_001069872.1 | 7 - 8 | 894 | 66 | |
| GenBank mRNA | BC116061.1 | - | 7 - 8 | 894 | 66 | |
More Information
Set Membership: |
|

BRMS1
Genomic Map