Assay Details
| Gene Symbol: MBOAT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075152.1 | NP_001068620.1 | 4 - 5 | 586 | 62 | |
| GenBank mRNA | BC119932.1 | - | 4 - 5 | 586 | 62 | |
More Information
Set Membership: |
|

MBOAT7
Genomic Map