Assay Details
| Gene Symbol: CCND2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076372.1 | NP_001069840.1 | 3 - 4 | 581 | 67 | |
| GenBank mRNA | BC120199.1 | - | 3 - 4 | 581 | 67 | |
More Information
Set Membership: |
|

CCND2
Genomic Map