Assay Details
| Gene Symbol: KBTBD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076463.1 | NP_001069931.1 | 3 - 4 | 759 | 131 | |
| GenBank mRNA | BC120403.1 | - | 3 - 4 | 759 | 131 | |
| BT030521.1 | - | 3 - 4 | 754 | 131 | ||
More Information
Set Membership: |
|

KBTBD4
Genomic Map