Assay Details
| Gene Symbol: ATP5F1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174684.2 | NP_777109.1 | 11 - 12 | 1601 | 86 | |
| GenBank mRNA | BC116059.1 | - | 11 - 12 | 86 | ||
| M22465.1 | - | 11 - 12 | 1601 | 86 | ||
More Information
Set Membership: |
|

ATP5F1A
Genomic Map