Assay Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100328.1 | NP_001093798.1 | 5 - 6 | 641 | 69 | |
| GenBank mRNA | BC148093.1 | - | 5 - 6 | 641 | 69 | |
More Information
Set Membership: |
|

OTULIN
Genomic Map