Assay Details
| Gene Symbol: IARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101069.2 | NP_001094539.1 | 1 - 2 | 60 | 94 | |
| GenBank mRNA | BC151484.1 | - | 1 - 2 | 61 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151484.1, NM_001101069.2 transcripts.
Set Membership: |
|

IARS1
Genomic Map