Assay Details
| Gene Symbol: AKAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102072.1 | NP_001095542.1 | 7 - 8 | 1094 | 61 | |
| GenBank mRNA | BC120047.1 | - | 7 - 8 | 1079 | 61 | |
| BC151317.1 | - | 7 - 8 | 1094 | 61 | ||
More Information
Set Membership: |
|

AKAP8
Genomic Map