Assay Details
| Gene Symbol: MPV17L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098145.2 | NP_001091614.1 | 3 - 4 | 458 | 86 | |
| GenBank mRNA | BC140467.1 | - | 3 - 4 | 459 | 86 | |
More Information
Set Membership: |
|

MPV17L2
Genomic Map