Assay Details
| Gene Symbol: KIF2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101147.1 | NP_001094617.1 | 5 - 6 | 542 | 55 | |
| GenBank mRNA | BC149279.1 | - | 5 - 6 | 542 | 55 | |
More Information
Set Membership: |
|

KIF2C
Genomic Map