Assay Details
| Gene Symbol: MAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105637.2 | NP_001099107.1 | 1 | 1650 | 108 | |
| GenBank mRNA | BC153214.1 | - | 1 | 1651 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105637.2 | NP_001099107.1 | 1 | 1650 | 108 | |
| GenBank mRNA | BC153214.1 | - | 1 | 1651 | 108 | |
Set Membership: |
|