Assay Details
| Gene Symbol: ADRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174231.1 | NP_776656.1 | 1 | 1948 | 135 | |
| GenBank mRNA | BC114132.1 | - | 1 | 1958 | 135 | |
| Z86037.1 | - | 1948 | 135 | |||
More Information
Set Membership: |
|

ADRB2
Genomic Map