Assay Details
| Gene Symbol: LOC789175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114862.1 | NP_001108334.1 | 1 - 2 | 84 | 91 | |
| GenBank mRNA | EU715239.2 | - | 1 - 2 | 135 | 91 | |
| KJ461433.1 | - | 2 - 3 | 311 | 91 | ||
More Information
Set Membership: |
|

LOC789175
Genomic Map