Assay Details
| Gene Symbol: MMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206637.1 | NP_001193566.1 | 1 - 2 | 157 | 76 | |
| GenBank mRNA | AB043995.1 | - | 1 - 2 | 102 | 76 | |
More Information
Set Membership: |
|

MMP3
Genomic Map