Assay Details
| Gene Symbol: INTS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101108.1 | NP_001094578.1 | 2 - 3 | 224 | 60 | |
| GenBank mRNA | BT030671.1 | - | 2 - 3 | 224 | 60 | |
More Information
Set Membership: |
|

INTS11
Genomic Map