Assay Details
| Gene Symbol: RCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103221.1 | NP_001096691.1 | 3 - 4 | 697 | 72 | |
| GenBank mRNA | BC134456.1 | - | 3 - 4 | 697 | 72 | |
More Information
Set Membership: |
|

RCN1
Genomic Map