Assay Details
| Gene Symbol: MAGED4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103311.1 | NP_001096781.1 | 6 - 7 | 1624 | 74 | |
| GenBank mRNA | BC147977.1 | - | 6 - 7 | 1624 | 74 | |
More Information
Set Membership: |
|

MAGED4B
Genomic Map