Assay Details
| Gene Symbol: MARCHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205812.3 | NP_001192741.1 | 19 - 20 | 1908 | 81 | |
| GenBank mRNA | BC119893.1 | - | 9 - 10 | 981 | 81 | |
More Information
Set Membership: |
|

MARCHF6
Genomic Map