Assay Details
| Gene Symbol: EEF2KMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163938.1 | NP_001157410.1 | 1 - 2 | 111 | 57 | |
| GenBank mRNA | BT025354.1 | - | 1 - 2 | 88 | 57 | |
| BT029841.1 | - | 1 - 2 | 96 | 57 | ||
More Information
Set Membership: |
|

EEF2KMT
Genomic Map