Assay Details
| Gene Symbol: PODN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192227.1 | NP_001179156.1 | 11 - 12 | 2020 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PODN| Gene Symbol: PODN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192227.1 | NP_001179156.1 | 11 - 12 | 2020 | 61 | |
Set Membership: |
|