Assay Details
| Gene Symbol: DDX3X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192962.1 | NP_001179891.1 | 15 - 16 | 1863 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX3X| Gene Symbol: DDX3X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192962.1 | NP_001179891.1 | 15 - 16 | 1863 | 64 | |
Set Membership: |
|