Assay Details
| Gene Symbol: C3H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038026.2 | NP_001033115.1 | 2 - 3 | 129 | 63 | |
| GenBank mRNA | BC109726.1 | - | 2 - 3 | 131 | 63 | |
More Information
Set Membership: |
|

C3H1orf189
Genomic Map