Assay Details
| Gene Symbol: MIR2368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030983.1 | 1 | 32 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-) | POMGNT2 |
AGO61, C22H3orf39, C3orf39, GTDC2 |
-174.0kb | Sense | Chr. 22 - 15060105 - 15080384 on NCBI Build ARS-UCD1.2 |
| Downstream | anoctamin 10 | ANO10 | TMEM16K | 53.0kb | Anti-sense | Chr. 22 - 15307508 - 15535485 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2368
Genomic Map