Assay Details
| Gene Symbol: MIR2452 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031193.1 | 1 | 41 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prosaposin like 1 | PSAPL1 | -183.5kb | Anti-sense | Chr. 6 - 113842038 - 113844510 on NCBI Build ARS-UCD1.2 | |
| Downstream | actin filament associated protein 1 | AFAP1 | 106.1kb | Anti-sense | Chr. 6 - 114134147 - 114283831 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2452
Genomic Map