Assay Details
| Gene Symbol: MIR6120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_107740.1 | 1 | 38 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | casein kinase 1 gamma 2 | CSNK1G2 | Sense | Chr. 7 - 44243766 - 44268033 on NCBI Build ARS-UCD1.2 | ||
| Downstream | sosondowah ankyrin repeat domain family member A | SOWAHA | ANKRD43 | 138.8kb | Sense | Chr. 7 - 44401108 - 44405294 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR6120
Genomic Map