Assay Details
| Gene Symbol: tns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318270.3 | NP_001305199.1 | 15 - 16 | 2853 | 85 | |
| NM_001318273.3 | NP_001305202.1 | 3 - 4 | 612 | 85 | ||
| NM_001383115.1 | 16 - 17 | 2937 | 85 | |||
| NM_059236.5 | NP_491637.2 | 17 - 18 | 2976 | 85 | ||
| NM_170867.5 | NP_740854.1 | 7 - 8 | 1873 | 85 | ||
More Information
Set Membership: |
|

tns-1
Genomic Map