Assay Details
| Gene Symbol: kin-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171029.8 | NP_741034.2 | 3 - 4 | 580 | 75 | |
| NM_171855.4 | NP_741035.2 | 4 - 5 | 760 | 75 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

kin-15| Gene Symbol: kin-15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171029.8 | NP_741034.2 | 3 - 4 | 580 | 75 | |
| NM_171855.4 | NP_741035.2 | 4 - 5 | 760 | 75 | ||
Set Membership: |
|