Assay Details
| Gene Symbol: tns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318270.3 | NP_001305199.1 | 23 - 24 | 3760 | 122 | |
| NM_001318273.3 | NP_001305202.1 | 11 - 12 | 1519 | 122 | ||
| NM_001318274.3 | NP_001305203.1 | 7 - 8 | 760 | 122 | ||
| NM_001318275.3 | NP_001305204.1 | 5 - 6 | 586 | 122 | ||
| NM_001383115.1 | 24 - 25 | 3844 | 122 | |||
| NM_059236.5 | NP_491637.2 | 25 - 26 | 3883 | 122 | ||
| NM_059237.7 | NP_491638.1 | 7 - 8 | 1349 | 122 | ||
More Information
Set Membership: |
|

tns-1
Genomic Map