Assay Details
| Gene Symbol: tns-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318270.3 | NP_001305199.1 | 3 - 4 | 392 | 101 | |
| NM_001383115.1 | 4 - 5 | 476 | 101 | |||
| NM_059236.5 | NP_491637.2 | 5 - 6 | 515 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383115.1 transcripts.
Set Membership: |
|

tns-1
Genomic Map