Assay Details
| Gene Symbol: eef-1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047635.6 | NP_001041100.1 | 5 - 6 | 1020 | 65 | |
| NM_001383394.1 | 5 - 6 | 1133 | 65 | |||
| NM_073399.6 | NP_505800.1 | 5 - 6 | 1210 | 65 | ||
More Information
Set Membership: |
|

eef-1G
Genomic Map