Assay Details
| Gene Symbol: slc-25A29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373160.1 | NP_001359577.1 | 7 - 8 | 1251 | 66 | |
| NM_182334.4 | NP_872134.1 | 6 - 7 | 1141 | 66 | ||
More Information
Set Membership: |
|

slc-25A29
Genomic Map