Assay Details
| Gene Symbol: vrk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062784.7 | NP_495185.1 | 5 - 6 | 1754 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

vrk-1| Gene Symbol: vrk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_062784.7 | NP_495185.1 | 5 - 6 | 1754 | 75 | |
Set Membership: |
|