Assay Details
| Gene Symbol: skpo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063367.7 | NP_495768.1 | 1 - 2 | 70 | 67 | |
| Gene Symbol: F14E5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026916.6 | NP_001022087.1 | 1 - 2 | 68 | 67 | |
| NM_001381525.2 | NP_001367096.1 | 1 - 2 | 71 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_063367.7 transcripts.
Set Membership: |
|

skpo-1
Genomic Map