Assay Details
| Gene Symbol: skpo-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063367.7 | NP_495768.1 | 6 - 7 | 1553 | 99 | |
| Gene Symbol: F14E5.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026916.6 | NP_001022087.1 | 6 - 7 | 1557 | 99 | |
| NM_001381525.2 | NP_001367096.1 | 6 - 7 | 1560 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_063367.7 transcripts.
Set Membership: |
|

skpo-1
Genomic Map