Assay Details
| Gene Symbol: dep-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267263.3 | NP_001254192.1 | 18 - 19 | 4568 | 94 | |
| NM_001267264.3 | NP_001254193.1 | 4 - 5 | 1104 | 94 | ||
More Information
Set Membership: |
|

dep-1
Genomic Map