Assay Details
| Gene Symbol: pir-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063558.6 | NP_495959.2 | 3 - 4 | 471 | 104 | |
| NR_023952.1 | 3 - 4 | 516 | 104 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pir-1| Gene Symbol: pir-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063558.6 | NP_495959.2 | 3 - 4 | 471 | 104 | |
| NR_023952.1 | 3 - 4 | 516 | 104 | |||
Set Membership: |
|