Assay Details
| Gene Symbol: cbp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129239.3 | NP_001122711.2 | 6 - 7 | 1695 | 83 | |
| NM_066759.5 | NP_499160.2 | 6 - 7 | 1734 | 83 | ||
| NM_066760.7 | NP_499161.2 | 6 - 7 | 1695 | 83 | ||
More Information
Set Membership: |
|

cbp-1
Genomic Map