Assay Details
| Gene Symbol: smc-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268189.3 | NP_001255118.1 | 17 - 18 | 3387 | 78 | |
| NM_001268190.3 | NP_001255119.1 | 4 - 5 | 728 | 78 | ||
More Information
Set Membership: |
|

smc-3
Genomic Map