Assay Details
| Gene Symbol: ssna-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028190.4 | NP_001023361.2 | 1 - 2 | 129 | 66 | |
| NM_001028191.8 | NP_001023362.1 | 3 - 4 | 1101 | 66 | ||
More Information
Set Membership: |
|

ssna-1
Genomic Map