Assay Details
| Gene Symbol: glt-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068809.9 | NP_501210.3 | 5 - 6 | 1370 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

glt-6| Gene Symbol: glt-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068809.9 | NP_501210.3 | 5 - 6 | 1370 | 64 | |
Set Membership: |
|