Assay Details
| Gene Symbol: F15E6.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068132.1 | NP_500533.1 | 1 | 448 | 76 | |
More Information
Set Membership: |
|
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

F15E6.5| Gene Symbol: F15E6.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068132.1 | NP_500533.1 | 1 | 448 | 76 | |
Set Membership: |
|
Genomic Map