Assay Details
| Gene Symbol: irld-44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071068.2 | NP_503469.2 | 4 - 5 | 1197 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

irld-44| Gene Symbol: irld-44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071068.2 | NP_503469.2 | 4 - 5 | 1197 | 140 | |
Set Membership: |
|