Assay Details
| Gene Symbol: gap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_076003.1 | NP_508404.1 | 9 - 10 | 1600 | 64 | |
| GenBank mRNA | AF118123.1 | - | 9 - 10 | 1600 | 64 | |
More Information
Set Membership: |
|

gap-1
Genomic Map