Assay Details
| Gene Symbol: sri-67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001269962.1 | NP_001256891.1 | 2 - 3 | 508 | 78 | |
| NM_001269963.1 | NP_001256892.1 | 2 - 3 | 508 | 78 | ||
More Information
Set Membership: |
|

sri-67
Genomic Map