Assay Details
| Gene Symbol: PDK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641208.3 | XP_005641265.1 | 3 - 4 | 508 | 98 | |
| XM_005641209.3 | XP_005641266.1 | 3 - 4 | 230 | 98 | ||
| XM_537984.6 | XP_537984.2 | 3 - 4 | 507 | 98 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PDK3
Genomic Map