Assay Details
| Gene Symbol: CALCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002948.2 | NP_001002948.1 | 2 - 3 | 193 | 77 | |
| XM_022407411.1 | XP_022263119.1 | 2 - 3 | 394 | 77 | ||
| GenBank mRNA | AB125102.1 | - | 2 - 3 | 194 | 77 | |
More Information
Set Membership: |
|

CALCB
Genomic Map