Assay Details
| Gene Symbol: IAPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003233.2 | NP_001003233.1 | 130 | 89 | ||
| GenBank mRNA | X59998.1 | - | 131 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: IAPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003233.2 | NP_001003233.1 | 130 | 89 | ||
| GenBank mRNA | X59998.1 | - | 131 | 89 | ||
Set Membership: |
|